Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Buffer Solutions

9,00, ±0.01, 25 °C, 4 L
  42421.K7
Buffer Solutions
  42421.K7
 :  
Buffer Solutions
Buffer Solutions

9,00, ±0.01, 25 °C, 4 L

 :  
 
1.130,00 kr
   
1 * 4 L
 

 

  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    9,00
  • Cat. No.:
    42421.K7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00147253