Buffer Solutions
6,86, ±0.01, 25 °C, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:6,86
- Cat. No.:42419.AP
- Physical form:Liquid
- MDL No.:MFCD00146206



