Buffer Solutions
3,00, ±0.01, 25 °C, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:3,00
- Cat. No.:42416.AP
- Physical form:Liquid
- MDL No.:MFCD00147253



