Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Funnel with short stem

75 mm, 9 mm, 75 mm
  221-1724
 :   Kavalier
Funnel with short stem
  221-1724
 :  Kavalier
Funnel with short stem
Funnel with short stem

75 mm, 9 mm, 75 mm

 
140,00 kr
   
1 * 1 Items
 

Borosilicate glass, grade 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With smooth internal walls.

 : DIN 12445 and ISO 4798

 
  • Funnel O-Ø:
    75 mm
  • Seq:
    09
  • Stem O-Ø:
    9 mm
  • Stem length:
    75 mm