Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Funnels, powder

120 mm, 30 mm, 30 mm
  221-1516
 :   Kavalier
Funnels, powder
  221-1516
 :  Kavalier
Funnels, powder
Funnels, powder

120 mm, 30 mm, 30 mm

 
407,00 kr
   
1 * 1 Items
 

Borosilicate glass 3,3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With smooth internal walls. For transferring powdery materials. Also suitable for use with liquids. Funnel rim is ground and fire-polished.

 
  • Funnel O-Ø:
    120 mm
  • Seq:
    07
  • Stem O-Ø:
    30 mm
  • Stem length:
    30 mm