Caps for tubes
Cap for 12 and 13 mm tubes, Natural
Supplier: DELTALAB
Jump to:
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Documents
Specifications
- Colour:Natural
- Description:Cap for 12 and 13 mm tubes