Order Entry
Denmark
Orders LinkContactUsLinkComponent
Laboratory trays
Laboratory trays
400×280×70 mm, 350×240×70 mm
Catalog # 216-1363
Supplier:   BOCHEM
Laboratory trays
Catalog # 216-1363
Supplier:  BOCHEM
Laboratory trays
Laboratory trays
400×280×70 mm, 350×240×70 mm
Price
1.160,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • Rounded corners for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, excellent thermal resistance
Multipurpose trays for a wide range of uses. Ideal as developing trays.
Specifications
  • W×D×H ext.:
    400×280×70 mm
  • Seq:
    05
  • W×D×H int.:
    350×240×70 mm