Order Entry
Denmark
ContactUsLinkComponent
Industrial storage containers hdpe, gray. affordable and versatile vessel. stackable with enough space for hubstaplerarme distance. suitable for liquids only up to about 50% level. 1 * 1 items
Catalog # 215-0258
Supplier:  SEMADENI
Industrial storage containers hdpe, gray. affordable and versatile vessel. stackable with enough space for hubstaplerarme distance. suitable for liquids only up to about 50% level. 1 * 1 items
Catalog # 215-0258
Supplier:  SEMADENI
Frequently Bought Together