Order Entry
Denmark
Orders LinkContactUsLinkComponent
Draining racks, wall mounted

Draining racks, wall mounted

Draining rack, with 75 rods of 100 mm, 450×600 mm
Catalog # 211-2084
Supplier:   SEMADENI
Draining racks, wall mounted
Catalog # 211-2084
Supplier:  SEMADENI
Draining racks, wall mounted

Draining rack, with 75 rods of 100 mm, 450×600 mm

Price
3.490,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

White, PVC drain channel, polyethylene coated metal rack.

  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning
Specifications
  • Description:
    Draining rack, with 75 rods of 100 mm
  • Seq:
    01
  • W×H:
    450×600 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.