Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffer Solutions

Buffer Solutions

2,00, ±0.01, 25 °C, 500 mL
Catalog # 40447.AP
Buffer Solutions
Catalog # 40447.AP
CAS Number:  
Buffer Solutions
Buffer Solutions

2,00, ±0.01, 25 °C, 500 mL

CAS Number:  
Price
397,00 kr
Log in for availability
1 * 500 mL
Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    2,00
  • Cat. No.:
    40447.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00146252
  • Appearance:
    Colourless

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.