Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffer Solutions

Buffer Solutions

4,01, ±0.01, 25 °C, 500 mL
Catalog # 38714.AP
Buffer Solutions
Catalog # 38714.AP
CAS Number:  
Buffer Solutions
Buffer Solutions

4,01, ±0.01, 25 °C, 500 mL

CAS Number:  
Price
400,00 kr
Log in for availability
1 * 500 mL
Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    4,01
  • Cat. No.:
    38714.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00146206

Specifications