Buffer Solutions
7,00, ±0.01, 25 °C, 4 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pk:4 L
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:7,00
- Cat. No.:38712.K7
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00146206
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



