Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Infrared optical windows

Crystal optic disc, polished both sides, Barium fluoride, 32 mm, 3 mm
  38436.KT
Infrared optical windows
  38436.KT
Infrared optical windows
Infrared optical windows

Crystal optic disc, polished both sides, Barium fluoride, 32 mm, 3 mm

 
6.930,00 kr
   
1 * 1 Items
 

These optical crystal windows are ideal for infrared spectroscopy.

  • Disc or rectangular
  • Polished or unpolished

A polishing kit is available for those who wish to polish their windows.

 
  • Window material:
    Barium fluoride
  • Description:
    Crystal optic disc, polished both sides
  • Dimensions:
    32 mm
  • Seq:
    42
  • Thickness:
    3 mm
Accessories

 

  •  
    Description
     
     
  • 39798.KT
    Crystal polishing kit
       
    1 * 1 Items - 7.940,00 kr