Infrared optical windows
Crystal optic disc, polished both sides, drilled, Sodium chloride, 32 mm, 3 mm
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
- Window material:Sodium chloride
- Description:Crystal optic disc, polished both sides, drilled
- Dimensions:32 mm
- Seq:88
- Thickness:3 mm



