Order Entry
Denmark
Orders LinkContactUsLinkComponent
Propylene oxide ≥99%
Propylene oxide ≥99%
Glass bottle, 1 L
  30765.K2
Propylene oxide ≥99%
  30765.K2
 :  75-56-9
Propylene oxide ≥99%
Propylene oxide ≥99%
Glass bottle, 1 L
 :  75-56-9
1.080,00 kr
1 * 1 L
Purity: 99+% CAS No.: 75-56-9 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Physical Form: Liquid Density: 0.83 Refractive Index: 1.3660 Appearance: Colourless Odour: Ether-like Melting Point: -112° Boiling Point: 34-35° Beilstein Reference: 79763 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00005126
 :  (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
  •  :75-56-9
  •  :C₃H₆O
  •  :MFCD00005126
  •  :58.08 g/mol
  •  :14,7856
  •  :34…35 °C
  •  :–112 °C
  •  :0.83
  •  :–37 °C (–35 °F)
  •  :Refrigerator
  •  :200-879-2
  •  :1280
  •  :3,I
  • Pack type:
    Glass bottle
  • Pk:
    1 L