Anthrone ACS
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Physical Form: Crystalline Sensitivity: Light Sensitive Melting Point: 155-158° Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187
Used for determination of carbohydrates
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Merck Index:14,691
- Melting Pt:155…158 °C
- Storage Temperature:Ambient
- EINECS:201-994-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



