Anthrone ACS
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Physical Form: Crystalline Sensitivity: Light Sensitive Melting Point: 155-158° Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187
Used for determination of carbohydrates
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Merck Index:14,691
- Melting Pt:155…158 °C
- Storage Temperature:Ambient
- EINECS:201-994-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



