Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acetone ≥99.5% semiconductor grade

Acetone ≥99.5% semiconductor grade

Glass bottle, 1 L
Catalog # 19392.K2
Flammable
Irritant
Acetone ≥99.5% semiconductor grade
Catalog # 19392.K2
CAS Number:  67-64-1
Acetone ≥99.5% semiconductor grade
Acetone ≥99.5% semiconductor grade

Glass bottle, 1 L

CAS Number:  67-64-1
Price
793,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 99.5% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 63580 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,66
  • Boiling Pt:56 °C
  • Melting Pt:–94 °C
  • Density:0.791
  • Flash Pt:–17 °C (1 °F)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications