Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

3-Methoxypropene ≥94%

Glass bottle for solids, 50 g
Catalog # 18652.18
Flammable
3-Methoxypropene ≥94%
Catalog # 18652.18
CAS Number:  627-40-7
3-Methoxypropene ≥94%
3-Methoxypropene ≥94%

Glass bottle for solids, 50 g

CAS Number:  627-40-7
Price
5.650,00 kr
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 94% CAS No.: 627-40-7 Molecular Formula: CH2=CHCH2OCH3 Formula Weight: 72.10 Flash Point: -23°(-94°F) Physical Form: Liquid Density: 0.77 Refractive Index: 1.3803 Odour: Ether-like Boiling Point: 42-43° UN No.: 3271 MDL No.: MFCD00008649

Synonyms:  Allyl methyl ether
  • CAS Number:627-40-7
  • Formula:C₄H₈O
  • MDL Number:MFCD00008649
  • Molecular Weight:72.11 g/mol
  • Boiling Pt:42…43 °C
  • Density:0.77
  • Flash Pt:–23 °C (–94 °F)
  • Storage Temperature:Ambient
  • EINECS:210-997-6
  • UN:3271
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications