Order Entry
Denmark
Orders LinkContactUsLinkComponent
3-Methoxypropene ≥94%
Glass bottle for solids, 10 g
  18652.09
3-Methoxypropene ≥94%
  18652.09
 :  627-40-7
3-Methoxypropene ≥94%
3-Methoxypropene ≥94%
Glass bottle for solids, 10 g
 :  627-40-7
1.270,00 kr
1 * 10 g
Purity: 94% CAS No.: 627-40-7 Molecular Formula: CH2=CHCH2OCH3 Formula Weight: 72.10 Flash Point: -23°(-94°F) Physical Form: Liquid Density: 0.77 Refractive Index: 1.3803 Odour: Ether-like Boiling Point: 42-43° UN No.: 3271 MDL No.: MFCD00008649
 :  Allyl methyl ether
  •  :627-40-7
  •  :C₄H₈O
  •  :MFCD00008649
  •  :72.11 g/mol
  •  :42…43 °C
  •  :0.77
  •  :–23 °C (–94 °F)
  •  :Ambient
  •  :210-997-6
  •  :3271
  •  :3,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g