Order Entry
Denmark
Orders LinkContactUsLinkComponent
n-Pentane ≥98%

n-Pentane ≥98%

Glass bottle, 1 L
Catalog # 17381.K2
Flammable
Health
Irritant
Environment
n-Pentane ≥98%
Catalog # 17381.K2
CAS Number:  109-66-0
n-Pentane ≥98%
n-Pentane ≥98%

Glass bottle, 1 L

CAS Number:  109-66-0
Price
895,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 98% CAS No.: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498

Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:14,7116
  • Boiling Pt:36 °C
  • Melting Pt:–130 °C
  • Density:0.626
  • Flash Pt:–49 °C (–57 °F)
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications