Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzo[a]pyrene ≥96%

Benzo[a]pyrene ≥96%

Glass bottle for solids, 100 mg
Catalog # 15856.MC
Health
Irritant
Benzo[a]pyrene ≥96%
Catalog # 15856.MC
CAS Number:  50-32-8
Benzo[a]pyrene ≥96%
Benzo[a]pyrene ≥96%

Glass bottle for solids, 100 mg

CAS Number:  50-32-8
Price
872,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents

Purity: 96% CAS No.: 50-32-8 Molecular Formula: C20H12 Formula Weight: 252.30 Physical Form: Crystalline Appearance: Pale yellow/green/orange Melting Point: 177-180° Boiling Point: 495° Solubility: Soluble in benzene, toluene, and xylene. Sparingly soluble in alcohol, methanol. Insoluble in water Merck Reference: 14,1103 UN No.: 3077 MDL No.: MFCD00003602

Extensively used in cancer research

Synonyms:  Benzo[def]chrysene, BaP
  • CAS Number:50-32-8
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00003602
  • Molecular Weight:252.32 g/mol
  • Merck Index:14,1103
  • Boiling Pt:495 °C
  • Melting Pt:177…180 °C
  • Density:1.351 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-028-5
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications