Benzo[a]pyrene ≥96%
Glass bottle for solids, 0,5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 50-32-8 Molecular Formula: C20H12 Formula Weight: 252.30 Physical Form: Crystalline Appearance: Pale yellow/green/orange Melting Point: 177-180° Boiling Point: 495° Solubility: Soluble in benzene, toluene, and xylene. Sparingly soluble in alcohol, methanol. Insoluble in water Merck Reference: 14,1103 UN No.: 3077 MDL No.: MFCD00003602
Extensively used in cancer research
Synonyms: Benzo[def]chrysene, BaP
- CAS Number:50-32-8
- Formula:C₂₀H₁₂
- MDL Number:MFCD00003602
- Molecular Weight:252.32 g/mol
- Merck Index:14,1103
- Boiling Pt:495 °C
- Melting Pt:177…180 °C
- Density:1.351 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-028-5
- UN:3077
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



