Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Plate stand, 200×316 mm, rod 16 mm Ø with height 800 mm, T 18 basic, T 25 digital, Plate stand R 1826, plate 200x316 mm, rod 800 mm
Catalog # 431-2901
Supplier: IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Catalog # 431-2901
Supplier: IKA
Jump to:
Product Details & Documents
Documents
VWR® for Stirring, Mixing, Shaking And Blending
Product Literature
Specifications
- Description:Plate stand, 200×316 mm, rod 16 mm Ø with height 800 mm
- Seq:104
- Type:Plate stand R 1826, plate 200x316 mm, rod 800 mm
- For:T 18 basic, T 25 digital
Accessories
Product Information
- Catalog #AvailabilityPrice
- Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mmStrap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm431-2944Log in for availability1 * 1 Items - kr3,570.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



