pH buffer solutions, referenced at 20 °C, Fluka™
13,00, ±0.02, Glycine, 20 °C, Plastic bottle, 1×1 L, 1 L
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33552-1L
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Glycine
- Pk:1 L
- Reference temperature:20 °C
- pH accuracy:±0.02
- Pk Info:1×1 L
- pH value:13,00



