Copper(I) cyanide
250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 544-92-3 Molecular Formula: CCuN Formula Weight: 89.56 Physical Form: Powder Density: 2.92 Appearance: White to cream Odour: Characteristic Melting Point: 473° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH Beilstein Reference: 3587244 Merck Reference: 14,2661 UN No.: 1587 MDL No.: MFCD00010975
Electroplating Cu or Fe, polymerization catalyst
Synonyms: Copper cyanide, Cuprous cyanide
- CAS Number:544-92-3
- Formula:CCuN
- MDL Number:MFCD00010975
- Molecular Weight:89.56 g/mol
- Merck Index:14,2661
- Melting Pt:473 °C
- Density:2.92
- Storage Temperature:Ambient
- EINECS:208-883-6
- UN:1587
- ADR:6.1,II
Documents
Specifications
Specifications
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



