Order Entry
Denmark
Orders LinkContactUsLinkComponent
Serological pipettes
Serological pipettes
2 ml, 0,01 ml, +, Green, 25/pack, 1000/case
Catalog # SARS86.1252.025
Marketsource
Supplier:   SARSTEDT
Serological pipettes
Catalog # SARS86.1252.025
Supplier:  SARSTEDT
Marketsource
Serological pipettes
2 ml, 0,01 ml, +, Green, 25/pack, 1000/case
Price
Inquire For Price
Log in for availability
1 * 1.000 Items
Product Details & Documents
PS, graduated, sterile.
  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging
Specifications
  • Colour code:
    Green
  • Packed:
    25/pack, 1000/case
  • Sterile:
    +
  • Capacity:
    2 ml
  • Division:
    0,01 ml