Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stand bases

Stand bases

125×200 mm, 1,25 kg, White
Catalog # 241-0177
Supplier:   R L ENTERPRISES
Stand bases
Catalog # 241-0177
Supplier:  R L ENTERPRISES
Stand bases
Stand bases

125×200 mm, 1,25 kg, White

Price
372,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Rectangular retort bases with M10 threaded hole in centre of short side.

  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing

Ordering information: Supplied without rods which need to be ordered separately.

Specifications
  • Colour:
    White
  • L×W:
    125×200 mm
  • Seq:
    03
  • Weight:
    1,25 kg