Double bosshead
Bosshead without blue plastic headed screw, Cast alloy, 16 mm
Catalog # RLAA2005/1PL
Supplier: R L ENTERPRISES
Jump to:
Product Details & Documents
Bossheads made from nickel plated cast alloy.
- Either with or without blue plastic headed nickel plated screws
- For use with rods up to 16 mm
Specifications
- Description:Bosshead without blue plastic headed screw
- Material:Cast alloy
- For rod Ø:16 mm