Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
Bosshead with blue plastic headed screw, Cast alloy, 16 mm
Catalog # 241-7867
Supplier:   R L ENTERPRISES
Double bosshead
Catalog # 241-7867
Supplier:  R L ENTERPRISES
Double bosshead
Double bosshead
Bosshead with blue plastic headed screw, Cast alloy, 16 mm
Price
244,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Bossheads made from nickel plated cast alloy.
  • Either with or without blue plastic headed nickel plated screws
  • For use with rods up to 16 mm
Specifications
  • Description:
    Bosshead with blue plastic headed screw
  • Material:
    Cast alloy
  • For rod Ø:
    16 mm