Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

High purity dNTPs

2'-Deoxythymidine 5'-Triphosphate, sodium (dTTP), 250 mg
Catalog # 27-1880-04
Supplier:   Cytiva
High purity dNTPs
Catalog # 27-1880-04
Supplier:  Cytiva
CAS Number:  
High purity dNTPs
High purity dNTPs

2'-Deoxythymidine 5'-Triphosphate, sodium (dTTP), 250 mg

CAS Number:  
Price
4.340,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents

dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy

Ordering information: Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.

Specifications

Specifications

  • Pk:
    250 mg
  • Description:
    2'-Deoxythymidine 5'-Triphosphate, sodium (dTTP)
  • Cat. No.:
    27-1880-04
  • Storage conditions:
    -20°C
  • Purity:
    ≥ 98% triphosphate

Specifications