Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
High purity dNTPs

High purity dNTPs

2'-Deoxyadenosine 5'-Triphosphate, disodium, crystalline (dATP), 250 mg
Catalog # 27-1850-04
Supplier:   Cytiva
High purity dNTPs
Catalog # 27-1850-04
Supplier:  Cytiva
CAS Number:  
High purity dNTPs
High purity dNTPs

2'-Deoxyadenosine 5'-Triphosphate, disodium, crystalline (dATP), 250 mg

CAS Number:  
Price
3.690,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents

dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy

Ordering information: Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.

Specifications

Specifications

  • Pakn:
    250 mg
  • Beskrivelse:
    2'-Deoxyadenosine 5'-Triphosphate, disodium, crystalline (dATP)
  • Cat. No.:
    27-1850-04
  • Renhed:
    ≥ 98% triphosphate
  • Opbevaring betingelser:
    -20°C

Specifications