Cork borer sets
Cork borers, set of 15, 5,0 - 22,0 mm
Catalog # 217-0508
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Brass, nickel plated, with handles and ejector.
- For manual boring of cork plugs
- Easy to clean
- Also available, cork drill sharpener compatible with all sets
Specifications
- Hole Ø:5,0 - 22,0 mm
- Description:Cork borers, set of 15
- Seq:05
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cork borer sharpener