Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cork borer sets
Cork borers, set of 6, 5,0 - 11,3 mm
Catalog # 235-0111
Supplier:   USBECK Laborgeräte
Cork borer sets
Catalog # 235-0111
Cork borer sets
Cork borer sets
Cork borers, set of 6, 5,0 - 11,3 mm
Price
504,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Brass, nickel plated, with handles and ejector.
  • For manual boring of cork plugs
  • Easy to clean
  • Also available, cork drill sharpener compatible with all sets
Specifications
  • Hole Ø:
    5,0 - 11,3 mm
  • Description:
    Cork borers, set of 6
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cork borer sharpener
    Cork borer sharpener
    217-9692
    Log in for availability
    1 * 1 Items - 343,00 kr