Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double bossheads
Double bossheads
With stainless steel T-screws, Stainless steel, electrolytically polished, 16,0 mm
Catalog # 241-7201
Supplier:   USBECK Laborgeräte
Double bossheads
Catalog # 241-7201
Double bossheads
Double bossheads
With stainless steel T-screws, Stainless steel, electrolytically polished, 16,0 mm
Price
394,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
For stand assembly.
  • Quick, easy assembly and disassembly
  • Robust, durable
  • Wide range of different materials available for many applications
Specifications
  • Description:
    With stainless steel T-screws
  • Material:
    Stainless steel, electrolytically polished
  • Seq:
    1
  • For rod Ø:
    16,0 mm