Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tripod stands

Tripod stands

Retort stand base, tripod, 1,0 kg, 115 mm
Catalog # 241-0245
Supplier:   USBECK Laborgeräte
Tripod stands
Catalog # 241-0245
Tripod stands
Tripod stands

Retort stand base, tripod, 1,0 kg, 115 mm

Price
294,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Steel, powder coated.

  • With M10 threaded hole
  • No support rod
  • Non-slip, rubber feet ensure firm footing
Specifications
  • Description:
    Retort stand base, tripod
  • Arm length:
    115 mm
  • Weight:
    1,0 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.