Order Entry
Denmark
Orders LinkContactUsLinkComponent
Burner, Teclu

Burner, Teclu

Teclu burner with air regulation needle valve, propane gas, tubing Ø 14 mm, Propane, 17 mm
Catalog # 451-0087
Supplier:   USBECK Laborgeräte
Burner, Teclu
Catalog # 451-0087
Burner, Teclu
Burner, Teclu

Teclu burner with air regulation needle valve, propane gas, tubing Ø 14 mm, Propane, 17 mm

Price
539,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Nickel plated brass base, in powder coated grey.

  • With air regulation valve and throttle screw
  • Without pilot light

Height: 165 mm

Specifications
  • Description:
    Teclu burner with air regulation needle valve, propane gas, tubing Ø 14 mm
  • Head Ø:
    17 mm
  • For:
    Propane
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.