SPECIAL KJELTABS AB03 1 * 1.000 Tablet
Catalog # THOMAB03
Supplier: THOMPSON & CAPPER
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Qualitative Filter Papers
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Sulphuric acid 95-97%, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Bathophenanthroline 99%
Supplier: Thermo Fisher Scientific