Containers with optional snap caps, Nunc™
for 12,5/20 and 30 ml containers, Natural
391-8579
Containers with optional snap caps, Nunc™
391-8579
PS, clear.
- Transparent polystyrene construction
- Versatile containers made of high-quality resins
- Snap caps available for all containers
- Cylindrical shape
- Non sterile
: Caps must be ordered seperately.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- Type:for 12,5/20 and 30 ml containers
- Cap colour:Natural
- Cat. No.:391-8579
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact