Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH indicator strips, Whatman™
pH indicator strips, Whatman™
Universal indicator papers, non bleeding strips, 0 - 14,0
Catalog # 700-0442
pH indicator strips, Whatman™
Catalog # 700-0442
pH indicator strips, Whatman™
pH indicator strips, Whatman™
Universal indicator papers, non bleeding strips, 0 - 14,0
Price
188,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
Specifications
  • pH range:
    0 - 14,0
  • Description:
    Universal indicator papers, non bleeding strips