Order Entry
Denmark
Orders LinkContactUsLinkComponent
Application specific filter papers, grade 287 ½, Whatman™

Application specific filter papers, grade 287 ½, Whatman™

287 ½, 185 mm
Katalog # 512-0582
Application specific filter papers, grade 287 ½, Whatman™
Katalog # 512-0582
Application specific filter papers, grade 287 ½, Whatman™
Application specific filter papers, grade 287 ½, Whatman™

287 ½, 185 mm

Price
722,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents

Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.

Specifikationer
  • Grade:
    287 ½
  • Seq:
    03
  • Size:
    185 mm
  • Thickness:
    360 µm
  • Cat. No.:
    512-0582
  • Filtration speed:
    330 s (Herzberg)
  • Weight:
    161 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.