Order Entry
Denmark
Orders LinkContactUsLinkComponent
Supelco® Reflectoquant® Test Strips, Peroxide
Supelco® Reflectoquant® Test Strips, Peroxide
Peroxide test, 0,2 - 20,0 mg/L H₂O₂, Enzymatic reaction, RQ, 50 Tests
Katalog # 1.16974.0001
Leverandør:   Merck
Supelco® Reflectoquant® Test Strips, Peroxide
Katalog # 1.16974.0001
Leverandør:  Merck
CAS nummer:  
Supelco® Reflectoquant® Test Strips, Peroxide
Supelco® Reflectoquant® Test Strips, Peroxide
Peroxide test, 0,2 - 20,0 mg/L H₂O₂, Enzymatic reaction, RQ, 50 Tests
CAS nummer:  
Price
485,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Test strips for the reflectometers RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
This test is suited for the determination of inorganic peroxides in aqueous solutions. Furthermore, it can be used in disinfecting and rinsing solutions (e.g., in food technology and laundries) and in swimming pool water.
Specifikationer
Specifikationer
  • Chemical method:
    Enzymatic reaction
  • Pk:
    50 Tests
  • Description:
    Peroxide test
  • Type:
    RQ
  • Range:
    0,2 - 20,0 mg/L H₂O₂
  • Cat. No.:
    1.16974.0001
  • M-Clarity quality level:
    MQ100
Specifikationer
Complementary Products
Produktinformation
  • Katalog #
    Description
    Model
    Tilgængelighed
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log ind for tilgængelighed
    1 * 1 Items - 10.580,00 kr