Test kits, ammonium, Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
ISO 23695: Water Quality — Determination of Ammonium Nitrogen in Water — Small-Scale Sealed Tube Method
• Method: This standard utilizes the Bertholet method (Indophenol blue) for determining ammonium.
• Citation Forms: Various forms such as NH₄⁺, NH₄-N, NH₃, NH₃-N are acceptable.
• Spectroquant® Cell Tests 1.14544, 1.14558, 1.14559, and 1.14739 correspond to this norm using 2-Chlorphenol.
Documents
Certifikater
Søg efter partinummerMerck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure Merck Segment Brochure
Product Literature
Test Kits, Ammonium, Spectroquant®, Supelco®
Additional Product Details
Specifikationer
Specifikationer
- Chemical method:Indophenol blue
- Pk:25 Tests
- Description:Ammonium cell test
- For unit:Prove, Nova 30/60, Multy, Move, Picco
- Range:4,0 - 80,0 mg/L NH₄ - N, 5,2 - 103,0 mg/L NH₄
- Cat. No.:1.14559.0001
- M-Clarity quality level:MQ100



