Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acrylamide, Millipore®

Acrylamide, Millipore®

Glass bottle, 1 kg
Catalog # 1.10784.1000
Toxic
Health
Supplier:   Merck
Acrylamide, Millipore®
Catalog # 1.10784.1000
Supplier:  Merck
CAS Number:  79-06-1
Acrylamide, Millipore®
Acrylamide, Millipore®

Glass bottle, 1 kg

CAS Number:  79-06-1
Price
2.320,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • ADR:6.1,
  •  :M-Clarity™ quality level = MQ200
  • UN:2074

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.