Sodium azide
Plastic bottle, 100 g
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:13,08653
- Melting Pt:275 °C
- Density:1.85 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Documents
Certificates
Search by Lot NumberMERC1.06688.0250_TDS_EN ProductPage_106688
Product specification
msds_106688
SDS
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



