Order Entry
Denmark
Orders LinkContactUsLinkComponent
On-column syringes

On-column syringes

General on-column syringe, stainless steel needle, 0,5 µl, 70 mm, 0,47 mm, 0.5BR-OC-7/0.47
Catalog # SGEA000376
On-column syringes
Catalog # SGEA000376
On-column syringes

General on-column syringe, stainless steel needle, 0,5 µl, 70 mm, 0,47 mm, 0.5BR-OC-7/0.47

Price
1.190,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

On-column syringes are suitable for general purpose applications.

When selecting an on-column syringe, the needle outside diameter (OD) must be smaller than the inside diameter (ID) of the capillary column it will be used with.

Specifications
  • Ext.Ø:
    0,47 mm
  • Type:
    0.5BR-OC-7/0.47
  • Volume:
    0,5 µl
  • Description:
    General on-column syringe, stainless steel needle
  • Length:
    70 mm