Order Entry
Denmark
Orders LinkContactUsLinkComponent
Colchicine

Colchicine

Glass bottle for solids, 1 g
Catalog # SERA77120.02
Toxic
Health
Marketsource
Supplier:   Serva
Colchicine
Catalog # SERA77120.02
Supplier:  Serva
Marketsource
CAS Number:  64-86-8
Colchicine

Glass bottle for solids, 1 g

CAS Number:  64-86-8
Price
380,85 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Melting Pt:154…156 °C
  • Density:1.32 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-598-5
  • UN:1544
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.