Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Histidine, Millipore®

L(+)-Histidine, Millipore®

Glass bottle, 100 g
Katalog # 1.04351.0100
Leverandør:   Merck
L(+)-Histidine, Millipore®
Katalog # 1.04351.0100
Leverandør:  Merck
CAS nummer:  71-00-1
L(+)-Histidine, Millipore®
L(+)-Histidine, Millipore®

Glass bottle, 100 g

CAS nummer:  71-00-1
Price
910,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents.

Synonymer:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS nummer:71-00-1
  • Formel:C₆H₉N₃O₂
  • MDL nummer:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Smeltende Pt:272…273 °C
  • Tæthed:1.44 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Ambient
  • EINECS:200-745-3
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.