Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Thiobarbituric acid

Glass bottle for solids, 10 g
Catalog # SERA36108.01
Marketsource
Supplier:   Serva
2-Thiobarbituric acid
Catalog # SERA36108.01
Supplier:  Serva
Marketsource
CAS Number:  504-17-6
2-Thiobarbituric acid

Glass bottle for solids, 10 g

CAS Number:  504-17-6
Price
215,37 kr
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  4,6-Dihydroxy-2-mercaptopyrimidine
  • CAS Number:504-17-6
  • Formula:C₄H₄N₂O₂S
  • MDL Number:MFCD00006674
  • Molecular Weight:144.15 g/mol
  • Boiling Pt:419 °C (1013 hPa)
  • Melting Pt:235 °C
  • Density:1.661 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-985-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications