D-(-)-Ribose
Glass bottle for solids, 10 g
Catalog # SERA34460.04
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:50-69-1
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00135453
- Molecular Weight:150.13 g/mol
- Merck Index:12,08371
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:90…95 °C
- Density:0.8 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-059-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



